MilliporeSigma
  • Antibody-conjugated soybean oil-filled calcium phosphate nanoshells for targetted delivery of hydrophobic molecules.
Antibody-conjugated soybean oil-filled calcium phosphate nanoshells for targetted delivery of hydrophobic molecules.
Journal of microencapsulation (2006-11-25)
H T Schmidt, M Kroczynski, J Maddox, Y Chen, R Josephs, A E Ostafin
ABSTRACT
Hollow calcium phosphate nanoparticles capable of encapsulating poorly water-soluble molecules were produced by self-assembly. Previously reported were solid calcium phosphate nanoparticles and water-filled calcium phosphate nanocapsules suited for encapsulating mostly hydrophilic, but not hydrophobic compounds. Here, calcium phosphate was deposited around 100 nm diameter, 1,2-dioleoyl-sn-glycero-3-phosphate stabilized soybean oil nanoemulsions using either calcium chloride or NaOH titrations to achieve shell thickness between 20-70 nm. The surface was functionalized with carboxylic acid via the addition of carboxyethylphosphonic acid to attach Molecular Probes AB-594C antibody using sulpho-n-hydroxysuccinimide and 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride with an efficiency of approximately 70%, while retaining near complete antibody function. Hydrophobic pyrene was encapsulated with an efficiency of 95%, at concentrations much higher than its water solubility limit, and exhibited spectral features characteristic of a hydrophobic environment. These materials can be used in the targeted delivery of many useful, yet poorly water-soluble pharmaceutical and nutraceutical compounds.
Sigma-Aldrich
3-Phosphonopropionic acid, technical grade, 94%