MilliporeSigma
  • Chromatographic behavior of the enantiomers of 2,2,2-trifluoro-1-(9-anthryl)ethanol on a quinidine-carbamate chiral stationary phase.
Chromatographic behavior of the enantiomers of 2,2,2-trifluoro-1-(9-anthryl)ethanol on a quinidine-carbamate chiral stationary phase.
Journal of chromatography. A (2006-01-06)
Leonid Asnin, Gustaf Götmar, Georges Guiochon
ABSTRACT
The enantioseparation of 2,2,2-trifluoro-1-(9-anthryl)ethanol on silica-bonded quinidine carbamate was examined under linear chromatographic conditions. The significant impact of nonselective adsorption on the retention was demonstrated. The influences of a polar additive in the mobile phase on the retention, the selectivity and the thermodynamic quantities of the retention were measured. A small effect of the pressure on the selectivity and on the accuracy of the thermodynamic measurements was observed.
Sigma-Aldrich
(R)-(−)-1-(9-Anthryl)-2,2,2-trifluoroethanol, ≥98%