MilliporeSigma
  • In vitro bulb development in shallot (Allium cepa L. Aggregatum Group): effects of anti-gibberellins, sucrose and light.
In vitro bulb development in shallot (Allium cepa L. Aggregatum Group): effects of anti-gibberellins, sucrose and light.
Annals of botany (2002-07-05)
F Le Guen-Le Saos, A Hourmant, F Esnault, J E Chauvin
ABSTRACT
Bulbing was studied in shallot plants cultured in vitro. Bulbing occurred under a 16 h photoperiod with fluorescent + incandescent light and 30-50 g 1(-1) sucrose in the culture medium. Exogenous gibberellin (10 microM GA3) inhibited leaf and root growth and bulbing. When added to the medium at a concentration of 10 microM, three inhibitors of gibberellin biosynthesis (ancymidol, flurprimidol and paclobutrazol) promoted bulb formation and the percentage of bulbing. When ancymidol was used in combination with GA3, it did not reverse the effect of GA3 applied alone. Under treatments with 30-70 g l(-1) sucrose, bulbing ratios greater than those found in control plants were achieved by addition of ancymidol, and bulb fresh weight was increased in the same way. Ancymidol caused a 66% decrease in sucrose content in leaf bases but greatly increased the glucose, fructose and fructan contents. The increase in fructan content by ancymidol could result from the three-fold rise in total [14C]sucrose uptake per plant from the culture medium associated with a marked increase in leaf base labelling at the expense of root labelling. The possible role of ancymidol is discussed and evidence supports a major regulatory role for gibberellins in bulbing.
Sigma-Aldrich
Ancymidol, suitable for plant cell culture, BioReagent