MilliporeSigma
  • Formal Synthesis of 4-diphosphocytidyl-2-C-methyl D-erythritol From D-(+)-Arabitol.
Formal Synthesis of 4-diphosphocytidyl-2-C-methyl D-erythritol From D-(+)-Arabitol.
Tetrahedron (2012-10-11)
Sina I Odejinmi, Rafael G Rascon, Wyman Chen, Kent Lai
ABSTRACT
2-C-methyl-D-erythritol-4-phosphate (MEP) is a key chemical intermediate of the non-mevalonate pathway for isoprenoid biosynthesis employed by many pathogenic microbes. MEP is also the precursor for the synthesis of 4-diphosphocytidyl-2-C-methyl D-erythritol (CDP-ME), another key intermediate of the non-mevalonate pathway. As this pathway is non-existent in higher animals, including humans, it represents great opportunities for novel antimicrobial development. To facilitate the in-depth studies of this pathway, we reported here a formal synthesis of CDP-ME through a new synthesis of 2-C-Methyl-D-erythritol-4-phosphoric acid from D-(+)-arabitol.
Sigma-Aldrich
2-C-Methyl-D-erythritol 4-phosphate lithium salt, ≥98% (TLC)