MilliporeSigma
  • Type II isopentenyl diphosphate isomerase: probing the mechanism with alkyne/allene diphosphate substrate analogues.
Type II isopentenyl diphosphate isomerase: probing the mechanism with alkyne/allene diphosphate substrate analogues.
Biochemistry (2010-06-22)
Nagendra K Sharma, Jian-Jung Pan, C Dale Poulter
ABSTRACT
Isopentenyl diphosphate isomerase (IDI) catalyzes the interconversion of isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP), the basic five-carbon building blocks of isoprenoid molecules. Two structurally unrelated classes of IDIs are known. Type I IPP isomerase (IDI-1) utilizes a divalent metal in a protonation-deprotonation reaction. In contrast, the type II enzyme (IDI-2) requires reduced flavin, raising the possibility that the reaction catalyzed by IDI-2 involves the net addition or abstraction of a hydrogen atom. As part of our studies of the mechanism of isomerization for IDI-2, we synthesized allene and alkyne substrate analogues for the enzyme. These molecules are predicted to be substantially less reactive toward proton addition than IPP and DMAPP but have similar reactivities toward hydrogen atom addition. This prediction was verified by calculations of gas-phase heats of reaction for addition of a proton and of a hydrogen atom to 1-butyne (3) and 1,2-butadiene (4) to form the 1-buten-2-yl carbocation and radical, respectively, and related affinities for 2-methyl-1-butene (5) and 2-methyl-2-butene (6) using G3MP2B3 and CBS-QB3 protocols. Alkyne 1-OPP and allene 2-OPP were not substrates for Thermus thermophilus IDI-2 or Escherichia coli IDI-1 but instead were competitive inhibitors. The experimental and computational results are consistent with a protonation-deprotonation mechanism for the enzyme-catalyzed isomerization of IPP and DMAPP.
Sigma-Aldrich
Isopentenyl pyrophosphate trilithium salt, ≥95.0% (TLC)