MilliporeSigma
  • Blue light irradiation inhibits the M2 polarization of the cancer-associated macrophages in colon cancer.
Blue light irradiation inhibits the M2 polarization of the cancer-associated macrophages in colon cancer.
BMC cancer (2024-06-01)
Toshiaki Yoshimoto, Masaaki Nishi, Shohei Okikawa, Kozo Yoshikawa, Takuya Tokunaga, Toshihiro Nakao, Chie Takasu, Hideya Kashihara, Yuma Wada, Takayuki Noma, Mitsuo Shimada
ABSTRACT
Recent studies have shown that blue light-emitting diode (LED) light has anti-tumor effects, suggesting the possibility of using visible light in cancer therapy. However, the effects of blue light irradiation on cells in the tumor microenvironment, including tumor-associated macrophages (TAMs), are unknown. Here, THP-1 cells were cultured in the conditioned medium (CM) of HCT-116 cells to prepare TAMs. TAMs were divided into LED-irradiated and control groups. Then, the effects of blue LED irradiation on TAM activation were examined. Expression levels of M2 macrophage markers CD163 and CD206 expression were significantly decreased in LED-irradiated TAMs compared with the control group. While control TAM-CM could induce HCT-116 cell migration, these effects were not observed in cells cultured in TAM-CM with LED irradiation. Vascular endothelial growth factor (VEGF) secretion was significantly suppressed in LED-exposed TAMs. PD-L1 expression was upregulated in HCT-116 cells cultured with TAM-CM but attenuated in cells cultured with LED-irradiated TAM-CM. In an in vivo model, protein expression levels of F4/80 and CD163, which are TAM markers, were reduced in the LED-exposed group. These results indicate that blue LED light may have an inhibitory effect on TAMs, as well as anti-tumor effects on colon cancer cells.
Sigma-Aldrich
Anti-OPN3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution