MilliporeSigma
  • Design of hepadnavirus core protein-based chimeric virus-like particles carrying epitopes from respiratory syncytial virus.
Design of hepadnavirus core protein-based chimeric virus-like particles carrying epitopes from respiratory syncytial virus.
NPJ vaccines (2024-03-20)
Shuai Shao, Xue Feng Zhang, Jun Wei Hou, Sen Sen Yang, Zi Bo Han, Hai Lan Wu, Fang Tang, Xin Yu Li, Ze Hua Lei, Zi Xin Zhao, Shu Xiang Li, Zhao Ming Liu, Pu Shan, Yu Qin Jin, Ji Guo Su, Yu Liang, Jing Zhang, Qi Ming Li
ABSTRACT
Respiratory syncytial virus (RSV) is one of the most important pathogens causing respiratory tract infection in humans, especially in infants and the elderly. The identification and structural resolution of the potent neutralizing epitopes on RSV fusion (F) protein enable an "epitope-focused" vaccine design. However, the display of RSV F epitope II on the surface of the widely-used human hepatitis B virus core antigen (HBcAg) has failed to induce neutralizing antibody response in mice. Here, we used the hepadnavirus core protein (HcAg) from different mammalian hosts as scaffolds to construct chimeric virus-like particles (VLPs) presenting the RSV F epitope II. Mouse immunization showed that different HcAg-based chimeric VLPs elicited significantly different neutralizing antibody responses, among which the HcAg derived from roundleaf bat (RBHcAg) is the most immunogenic. Furthermore, RBHcAg was used as the scaffold platform to present multiple RSV F epitopes, and the immunogenicity was further improved in comparison to that displaying a single epitope II. The designed RBHcAg-based multiple-epitope-presenting VLP formulated with MF59-like adjuvant elicited a potent and balanced Th1/Th2 immune response, and offered substantial protection in mice against the challenge of live RSV A2 virus. The designed chimeric VLPs may serve as the potential starting point for developing epitope-focused vaccines against RSV. Our study also demonstrated that RBHcAg is an effective VLP carrier for presenting foreign epitopes, providing a promising platform for epitope-focused vaccine design.
Sigma-Aldrich
Anti-RSV Antibody, nucleoprotein, clone 130-12H, clone 130-12H, Chemicon®, from mouse