MilliporeSigma
  • Endothelial DR6 in blood-brain barrier malfunction in Alzheimer's disease.
Endothelial DR6 in blood-brain barrier malfunction in Alzheimer's disease.
Cell death & disease (2024-04-13)
Xiaomin Huang, Junhua Qi, Yixun Su, Ying Zhou, Qi Wang, Taida Huang, Dongdong Xue, Yunxin Zeng, Alexei Verkhratsky, Benjie Zhou, Hui Chen, Chenju Yi
ABSTRACT
The impairment of the blood-brain barrier (BBB) has been increasingly recognised as a critical element in the early pathogenesis of Alzheimer's disease (AD), prompting a focus on brain endothelial cells (BECs), which serve as the primary constituents of the BBB. Death receptor 6 (DR6) is highly expressed in brain vasculature and acts downstream of the Wnt/β-catenin pathway to promote BBB formation during development. Here, we found that brain endothelial DR6 levels were significantly reduced in a murine model of AD (APPswe/PS1dE9 mice) at the onset of amyloid-β (Aβ) accumulation. Toxic Aβ25-35 oligomer treatment recapitulated the reduced DR6 in cultured BECs. We further showed that suppressing DR6 resulted in BBB malfunction in the presence of Aβ25-35 oligomers. In contrast, overexpressing DR6 increased the level of BBB functional proteins through the activation of the Wnt/β-catenin and JNK pathways. More importantly, DR6 overexpression in BECs was sufficient to rescue BBB dysfunction in vitro. In conclusion, our findings provide new insight into the role of endothelial DR6 in AD pathogenesis, highlighting its potential as a therapeutic target to tackle BBB dysfunction in early-stage AD progression.
Sigma-Aldrich
Anti-SLC2A1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution