MilliporeSigma
  • Unveiling the Regulatory Role of SIRT1 in Oxidative Stress Response of bovine mammary cells.
Unveiling the Regulatory Role of SIRT1 in Oxidative Stress Response of bovine mammary cells.
Journal of dairy science (2024-06-15)
Yufei Zhang, Juxiong Liu, Shuai Yuan, Shu Liu, Meng Zhang, Huijie Hu, Yu Cao, Guiqiu Hu, Shoupeng Fu, Wenjin Guo
ABSTRACT
High-yield dairy cows typically undergo intense cellular metabolism, leading to oxidative stress in their mammary tissues. Our study found that these high-yield cows had significantly elevated levels of hydrogen peroxide (H2O2), lipoperoxidase, and total antioxidant capacity in their blood, compared with ordinary cows. This increased oxidative stress is associated with heightened expression of genes such as GCLC, GCLM and SIRT1 and proteins such as SIRT1 in the mammary tissue of high-yield cows. MAC-T cells were stimulated with H2O2 at a concentration equal to the average H2O2 level in the serum of ethically high-yielding cows, as detected by an assay kit. Our observations revealed that short-term exposure (12 h) to H2O2 upregulated the expression of SIRT1 gene and protein. It also increased gene expression for SOD2, CAT, GCLC, GCLM, PGC-1α, and NQO1, elevated the phosphorylation of AMPK, and enhanced protein expression of PGC-1α, NQO1, Nrf2, and HO-1, while reducing the phosphorylation of NF-κB. Additionally, short-term H2O2 stimulation resulted in increased total antioxidant capacity, SOD, GSH, and CAT levels in the mammary epithelial cells of dairy cows. In contrast, prolonged exposure to H2O2 (24 h) yielded opposite results, indicating reduced antioxidant capacity. Further investigation showed that SIRT1 inhibitor (EX 527) could reverse the enhanced cellular antioxidant capacity triggered by short-term oxidative stress. However, it is crucial to note that while 12 h H2O2 stimulation improved antioxidant capacity, reactive oxygen species (ROS) and malondialdehyde (MDA) levels inside the cell gradually increased over time, suggesting greater damage under long-term stimulation. Conversely, the SIRT1 activator (SRT 2104) could reverse the reduced cellular antioxidant capacity caused by long-term oxidative stress and significantly inhibit the accumulation of ROS and MDA. Notably, SRT 2104 demonstrated similar effects in MAC-T cells during lactation. In summary, SIRT1 plays a crucial role in regulating the antioxidant capacity of mammary epithelial cells in dairy cows. This discovery provides valuable insights into the antioxidant mechanisms of mammary cells, which can serve as a theoretical foundation for future mammary health strategies.
Product Description
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard