MilliporeSigma
  • Site-Specific Conjugation of Native Antibodies Using Engineered Microbial Transglutaminases.
Site-Specific Conjugation of Native Antibodies Using Engineered Microbial Transglutaminases.
Bioconjugate chemistry (2020-03-07)
Stephan Dickgiesser, Marcel Rieker, Dirk Mueller-Pompalla, Christian Schröter, Jason Tonillo, Shira Warszawski, Sabine Raab-Westphal, Stefanie Kühn, Tim Knehans, Doreen Könning, Julia Dotterweich, Ulrich A K Betz, Jan Anderl, Stefan Hecht, Nicolas Rasche
ABSTRACT
Site-specific bioconjugation technologies are frequently employed to generate homogeneous antibody-drug conjugates (ADCs) and are generally considered superior to stochastic approaches like lysine coupling. However, most of the technologies developed so far require undesired manipulation of the antibody sequence or its glycan structures. Herein, we report the successful engineering of microbial transglutaminase enabling efficient, site-specific conjugation of drug-linker constructs to position HC-Q295 of native, fully glycosylated IgG-type antibodies. ADCs generated via this approach demonstrate excellent stability in vitro as well as strong efficacy in vitro and in vivo. As it employs different drug-linker structures and several native antibodies, our study additionally proves the broad applicability of this approach.
Sigma-Aldrich
Enhanced Microbial Transglutaminase, Glycosylation tolerant, for site-specific antibody bioconjugation