MilliporeSigma
  • Effects of green light-emitting diode irradiation on hepatic differentiation of hepatocyte-like cells generated from human adipose-derived mesenchymal cells.
Effects of green light-emitting diode irradiation on hepatic differentiation of hepatocyte-like cells generated from human adipose-derived mesenchymal cells.
Scientific reports (2023-11-16)
Yuhei Waki, Yu Saito, Shuhai Chen, Tetsuya Ikemoto, Takayuki Noma, Hiroki Teraoku, Shinichiro Yamada, Yuji Morine, Mitsuo Shimada
ABSTRACT
Light-emitting diode (LED) irradiation has been used in the differentiation of mesenchymal stem cells into a variety of cell types. This study investigated the effect of green LED (GLED) irradiation on the differentiation of adipocyte-derived mesenchymal cells into hepatocyte-like cells (HLCs) and the mechanism of its action. HLCs in the hepatocyte maturation phase were irradiated with GLED (520 nm, 21 W/m2, 5 min/day for 10 days). The cells were then assessed for expression of hepatocyte maturity genes and opsin 3 (OPN3), hepatocyte function, viability, apoptosis, and levels of reactive oxygen species (ROS), intracellular adenosine triphosphate (ATP) and calcium ions (Ca2+). GLED irradiation increased Alpha-1 antitrypsin and Ornithine transcarbamylase gene expression, promoted Cytochrome P450 3A4 activity and urea synthesis, and elevated intracellular ROS, ATP and Ca2+ levels. OPN3 expression was significantly more upregulated in GLED-irradiated HLCs than in the non-irradiated HLCs. No significant difference in cell viability or apoptosis was observed between GLED-irradiated and non-irradiated HLCs. GLED irradiation can promote hepatocyte maturation and functions through OPN3. GLED irradiation also stimulated mitochondrial function via Ca2+/ATP/ROS activation. GLED irradiation has potential to support cell-based transplantation in patients.
Sigma-Aldrich
Anti-OPN3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution