MilliporeSigma
  • Hypothesis: Metabolic targeting of 5-aminolevulinate synthase by tryptophan and inhibitors of heme utilisation by tryptophan 2,3-dioxygenase as potential therapies of acute hepatic porphyrias.
Hypothesis: Metabolic targeting of 5-aminolevulinate synthase by tryptophan and inhibitors of heme utilisation by tryptophan 2,3-dioxygenase as potential therapies of acute hepatic porphyrias.
Medical hypotheses (2019-08-25)
Abdulla A-B Badawy
ABSTRACT
Metabolic targeting of liver 5-aminolevulinate synthase (5-ALAS) by inhibition of heme utilisation by tryptophan (Trp) 2,3-dioxygenase (TDO) or the use of tryptophan is proposed as a therapy of acute hepatic porphyrias. 5-ALAS, the rate-limiting enzyme of heme biosynthesis, is under negative feedback control by a small regulatory heme pool in the hepatic cytosol. Acute porphyric attacks, precipitated by fasting, certain hormones and some drugs, involve induction of 5-ALAS secondarily to depletion of the above pool, and the resultant elevation of 5-ALA levels initiates the abdominal and neurological symptoms of attacks. By utilising the regulatory heme, cytosolic TDO undermines the feedback control, thus allowing 5-ALAS induction to occur, e.g. upon glucocorticoid induction of TDO during fasting (starvation) and exogenous glucocorticoid administration. Currently, glucose therapy is the preferred strategy for reversing moderate attacks induced by fasting (calorie restriction), with more severe attacks being treated by intravenous heme preparations. Reversal of fasting-induced attacks by glucose is explained by the previously demonstrated reversal of increased heme utilisation by TDO. Inhibitors of this utilisation are therefore potential therapeutic targets in acute attacks and also for maintenance of a symptomless state. Existing TDO inhibitors other than glucose include allopurinol, nicotinamide and recently developed potent inhibitors such as LM10 used in cancer therapy. Based on studies in rats, the hypothesis predicts that the safety or otherwise of drugs in the hepatic porphyrias is determined by their ability to inhibit TDO utilisation of heme under basal conditions or after glucocorticoid induction or heme activation of TDO, in parallel with reciprocal changes in 5-ALAS induction. Tryptophan is also proposed as a potential therapy of acute attacks either alone or as an adjunct to the recently proposed 5-ALAS1 gene silencing. Trp increases heme biosynthesis by enhancing 5-ALA dehydratase activity and, based on a Trp-5-ALA model presented herein, Trp offers several advantages over heme therapy, namely rapid conversion of 5-ALA into heme, a greatly enhanced heme availability, a near complete inhibition of 5-ALAS induction, assumed rapid clearance of 5-ALA and hence accelerated resolution of symptoms of attacks, and finally provision of the neuroprotective metabolite kynurenic acid to neutralise the neurological symptoms. The hypothesis also addresses heme regulation in species lacking the TDO free apoenzyme and its glucocorticoid induction mechanism and proposes detailed assessment of heme biosynthesis in these species. Detailed proposals for testing the hypothesis are presented.
Sigma-Aldrich
LM10, ≥98% (HPLC)