MilliporeSigma
  • Visualizing inflammation with an M1 macrophage selective probe via GLUT1 as the gating target.
Visualizing inflammation with an M1 macrophage selective probe via GLUT1 as the gating target.
Nature communications (2022-10-11)
Heewon Cho, Haw-Young Kwon, Amit Sharma, Sun Hyeok Lee, Xiao Liu, Naoki Miyamoto, Jong-Jin Kim, Sin-Hyeog Im, Nam-Young Kang, Young-Tae Chang
ABSTRACT
Macrophages play crucial roles in protecting our bodies from infection and cancers. As macrophages are multi-functional immune cells, they have diverse plastic subsets, such as M1 and M2, derived from naïve M0 cells. Subset-specific macrophage probes are essential for deciphering and monitoring the various activation of macrophages, but developing such probes has been challenging. Here we report a fluorescent probe, CDr17, which is selective for M1 macrophages over M2 or M0. The selective staining mechanism of CDr17 is explicated as Gating-Oriented Live-cell Distinction (GOLD) through overexpressed GLUT1 in M1 macrophages. Finally, we demonstrate the suitability of CDr17 to track M1 macrophages in vivo in a rheumatoid arthritis animal model.
Sigma-Aldrich
Anti-GLUT-1 antibody, Rabbit monoclonal, clone SP168, recombinant, expressed in proprietary host, affinity isolated antibody