MilliporeSigma
  • WTAP-mediated N6-methyladenosine modification of NLRP3 mRNA in kidney injury of diabetic nephropathy.
WTAP-mediated N6-methyladenosine modification of NLRP3 mRNA in kidney injury of diabetic nephropathy.
Cellular & molecular biology letters (2022-06-28)
Jianzi Lan, Bowen Xu, Xin Shi, Qi Pan, Qing Tao
ABSTRACT
Diabetic nephropathy (DN) is prevalent in patients with diabetes. N6-methyladenosine (m6A) methylation has been found to cause modification of nucleotide-binding oligomerization domain, leucine-rich repeat, and pyrin domain-containing (NLRP) 3, which is involved in cell pyroptosis and inflammation. WTAP is a key gene in modulating NLRP3 m6A. In this study, WTAP was silenced or overexpressed in high glucose (HG)-treated HK-2 cells to determine its influence on pyroptosis, NLRP3 inflammasome-related proteins, and the release of pro-inflammatory cytokines. NLRP3 expression and m6A levels were assessed in the presence of WTAP shRNA (shWTAP). WTAP expression in HK-2 cells was examined with the introduction of C646, a histone acetyltransferase p300 inhibitor. We found that WTAP expression was enhanced in patients with DN and in HG-treated HK-2 cells. Knockdown of WTAP attenuated HG-induced cell pyroptosis and NLRP3-related pro-inflammatory cytokines in both HK-2 cells and db/db mice, whereas WTAP overexpression promoted these cellular processes in HK-2 cells. WTAP mediated the m6A of NLRP3 mRNA that was stabilized by insulin-like growth factor 2 mRNA binding protein 1. Histone acetyltransferase p300 regulated WTAP expression. WTAP mRNA levels were positively correlated with NLRP3 inflammasome components and pro-inflammatory cytokines. Taken together, WTAP promotes the m6A methylation of NLRP3 mRNA to upregulate NLRP3 inflammasome activation, which further induces cell pyroptosis and inflammation.
Sigma-Aldrich
GenElute mRNA Miniprep Kit, sufficient for 10 purifications