MilliporeSigma
  • LRPPRC regulates redox homeostasis via the circANKHD1/FOXM1 axis to enhance bladder urothelial carcinoma tumorigenesis.
LRPPRC regulates redox homeostasis via the circANKHD1/FOXM1 axis to enhance bladder urothelial carcinoma tumorigenesis.
Redox biology (2021-12-06)
Wen-Su Wei, Ning Wang, Min-Hua Deng, Pei Dong, Jian-Ye Liu, Zhen Xiang, Xiang-Dong Li, Zhi-Yong Li, Zhen-Hua Liu, Yu-Lu Peng, Zhen Li, Li-Juan Jiang, Kai Yao, Yun-Lin Ye, Wen-Hua Lu, Zhi-Ling Zhang, Fang-Jian Zhou, Zhuo-Wei Liu, Dan Xie, Chun-Ping Yu
ABSTRACT
Reactive oxygen species (ROS) which are continuously generated mainly by mitochondria, have been proved to play an important role in the stress signaling of cancer cells. Moreover, pentatricopeptide repeat (PPR) proteins have been suggested to take part in mitochondrial metabolism. However, the mechanisms integrating the actions of these distinct networks in urothelial carcinoma of the bladder (UCB) pathogenesis are elusive. In this study, we found that leucine rich pentatricopeptide repeat containing (LRPPRC) was frequently upregulated in UCB and that it was an independent prognostic factor in UCB. We further revealed that LRPPRC promoted UCB tumorigenesis by regulating the intracellular ROS homeostasis. Mechanistically, LRPPRC modulates ROS balance and protects UCB cells from oxidative stress via mt-mRNA metabolism and the circANKHD1/FOXM1 axis. In addition, the SRA stem-loop interacting RNA binding protein (SLIRP) directly interacted with LRPPRC to protect it from ubiquitination and proteasomal degradation. Notably, we showed that LRPPRC modulated the tumorigenesis of UCB cells in a circANKHD1-FOXM1-dependent manner. In conclusion, LRPPRC exerts critical roles in regulating UCB redox homeostasis and tumorigenesis, and is a prognostic factor for UCB; suggesting that LRPPRC may serve as an exploitable therapeutic target in UCB.
Sigma-Aldrich
Anti-Superoxide Dismutase (MnSOD) (DD-17) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution