MilliporeSigma
  • Purification and characterization of a carboxypeptidase from squid hepatopancreas (Illex illecebrosus).
Purification and characterization of a carboxypeptidase from squid hepatopancreas (Illex illecebrosus).
Journal of agricultural and food chemistry (2001-10-16)
R Raksakulthai, N F Haard
ABSTRACT
The hepatopancreas of squid (Illex illecebrosus) extract contains a wide range of carboxypeptidase (CP) activities based on hydrolysis of N-CBZ-dipeptide substrates. SDS-PAGE zymograms with N-CBZ-Phe-Leu substrate revealed three activity zones (CP-I, 23 kDa; CP-II, 29 kDa; CP-III, 42 kDa). CP-I was purified 225-fold with 86.20% recovery based on N-CBZ-Ala-Phe activity by chromatography on DEAE-cellulose, gel filtration, and chromatofocusing. The purified enzyme had broad specificity toward N-CBZ-dipeptides; however, it preferred substrates with a hydrophobic amino acid at the C terminus. CP-I had greatest activity with N-CBZ-Ala-Phe (specific activity = 7104 units/mg of protein, K(m) = 0.40 mM, and physiological efficiency = 22863). CP-I had a pI of 3.4 and is a metalloprotease that is activated by Co(2+) and partially inhibited by Pefabloc, a serine protease inhibitor. With N-CBZ-Ala-Phe and Gly-Phe, it had optimum activity at pH 8 and 70 degrees C. The amino acid composition of squid CP-I is similar to that of CP A from other species.
Sigma-Aldrich
Z-Phe-Leu