MilliporeSigma
  • Cross-reactivity of IgM anti-modified protein antibodies in rheumatoid arthritis despite limited mutational load.
Cross-reactivity of IgM anti-modified protein antibodies in rheumatoid arthritis despite limited mutational load.
Arthritis research & therapy (2021-09-05)
Sanne Reijm, Theresa Kissel, Gerrie Stoeken-Rijsbergen, Linda M Slot, Corrie M Wortel, Hugo J van Dooren, Nivine E W Levarht, Arieke S B Kampstra, Veerle F A M Derksen, Pleuni Ooijevaar-de Heer, Holger Bang, Jan W Drijfhout, Leendert A Trouw, Tom W J Huizinga, Theo Rispens, Hans U Scherer, René E M Toes
ABSTRACT
Anti-modified protein antibodies (AMPA) targeting citrullinated, acetylated and/or carbamylated self-antigens are hallmarks of rheumatoid arthritis (RA). Although AMPA-IgG cross-reactivity to multiple post-translational modifications (PTMs) is evident, it is unknown whether the first responding B cells, expressing IgM, display similar characteristics or if cross-reactivity is crucially dependent on somatic hypermutation (SHM). We now studied the reactivity of (germline) AMPA-IgM to further understand the breach of B cell tolerance and to identify if cross-reactivity depends on extensive SHM. Moreover, we investigated whether AMPA-IgM can efficiently recruit immune effector mechanisms. Polyclonal AMPA-IgM were isolated from RA patients and assessed for cross-reactivity towards PTM antigens. AMPA-IgM B cell receptor sequences were obtained by single cell isolation using antigen-specific tetramers. Subsequently, pentameric monoclonal AMPA-IgM, their germline counterparts and monomeric IgG variants were generated. The antibodies were analysed on a panel of PTM antigens and tested for complement activation. Pentameric monoclonal and polyclonal AMPA-IgM displayed cross-reactivity to multiple antigens and different PTMs. PTM antigen recognition was still present, although reduced, after reverting the IgM into germline. Valency of AMPA-IgM was crucial for antigen recognition as PTM-reactivity significantly decreased when AMPA-IgM were expressed as IgG. Furthermore, AMPA-IgM was 15- to 30-fold more potent in complement-activation compared to AMPA-IgG. We provide first evidence that AMPA-IgM are cross-reactive towards different PTMs, indicating that PTM (cross-)reactivity is not confined to IgG and does not necessarily depend on extensive somatic hypermutation. Moreover, our data indicate that a diverse set of PTM antigens could be involved in the initial tolerance breach in RA and suggest that AMPA-IgM can induce complement-activation and thereby inflammation.
Sigma-Aldrich
Goat Anti-Human IgM Antibody, Fc5µ, HRP conjugate, Chemicon®, from goat