MilliporeSigma
  • Murine VCAM-1. Molecular cloning, mapping, and analysis of a truncated form.
Murine VCAM-1. Molecular cloning, mapping, and analysis of a truncated form.
Journal of immunology (Baltimore, Md. : 1950) (1994-11-01)
A G Kumar, X Y Dai, C A Kozak, M P Mims, A M Gotto, C M Ballantyne
ABSTRACT
Vascular cell adhesion molecule-1 (VCAM-1) is a member of the Ig superfamily that shows increased expression in a number of pathologic conditions. The role of VCAM-1 in human disease remains undefined and murine models are being extensively studied to help define the importance of VCAM-1 in inflammatory disorders. We have cloned and characterized the murine Vcam1 gene including 3 kb of 5'-flanking sequences and mapped the gene to chromosome 3 near Amy1. cDNA clones isolated from a stimulated hepatic library were found to encode a truncated form of VCAM-1 (T-VCAM-1) which contains Ig domains 1 through 3 and has a unique alternative carboxyl terminus. This form arises by alternative splicing. High level expression of T-VCAM-1 in transfected L cells was sufficient to support adhesion of lymphocytes, and this adhesion was blocked by Abs to VCAM-1. Treatment of transfected COS cells with phospholipase C led to reduced levels of T-VCAM-1 on the cell surface consistent with glycosylphosphatidylinositol linkage. Northern blot analysis showed that mRNA for T-VCAM-1 is inducible in multiple tissues after stimulation with endotoxin. Both forms of VCAM-1 were expressed in cultured endothelial, fibroblast, and aortic smooth muscle cells, whereas neither form was observed in monocyte- and lymphocyte-derived lines. Differential regulation of both forms of VCAM-1 was observed in the three different cell types that are present in the vessel wall. Thus, expression of VCAM-1 is restricted and controlled at the level of transcription and by alternative splicing.