MilliporeSigma
  • Characterizing the Low-Dose Effects of Methylmercury on the Early Stages of Embryo Development Using Cultured Human Embryonic Stem Cells.
Characterizing the Low-Dose Effects of Methylmercury on the Early Stages of Embryo Development Using Cultured Human Embryonic Stem Cells.
Environmental health perspectives (2021-07-31)
Bai Li, Cunye Qiao, Xiaolei Jin, Hing Man Chan
ABSTRACT
Global concerns of methylmercury (MeHg) exposure have been raised, especially on its effects on pregnant women. Recent epidemiological studies have revealed associations between maternal blood/hair MeHg concentrations, adverse pregnancy outcomes, and developmental deficits. However, the underlying mechanisms remain unclear. In this study, we characterized the effects of MeHg exposure on undifferentiated human embryonic stem cells (hESCs) and extrapolated the effects to human embryonic development. hESCs were exposed to 0, 1, 5, 10, 50, 100 or 200nM MeHg for 24 h or 6 d. Cell adherence and colony formation and expansion were examined under the microscope. Cell attachment, viability/proliferation, apoptosis, stress response, cell cycle, autophagy, and expression of cell lineage marker genes and proteins were measured at the end of exposures. Our results indicated that exposure to nanomolar concentrations of MeHg was associated with a) higher levels of reactive oxygen species (ROS) and hemeoxygenase-1 (HO-1), suggesting increased stress and adaptive responses; b) lower cellular adhesion, viability/proliferation, and colony formation and expansion; c) higher levels of apoptosis, reflected by higher cleaved caspase-3 expression and Annexin V binding; d) higher levels of cytoskeleton protein α-tubulin expression; e) higher rates of G1/S phase cell cycle arrest; and f) autophagy inhibition, as shown by a lower LC3BII/LC3BI ratio and accumulation of SQSTM1 (p62). These outcomes were accompanied by higher expressions of self-renewal genes or proteins or both, including OCT4, SOX2, NANOG, and cytokine receptor IL6ST, as well as pluripotency and the cell fate regulator cyclin D1. These results revealed that under the selection pressure of exposure to low doses of MeHg, some hESCs underwent apoptosis, whereas others adapted and survived with enhanced self-renewal gene expression and specific morphological phenotypes. Findings from the present study provide in vitro evidence that low doses of MeHg adversely affect hESCs when exposed during a period of time that models embryonic pre-, during, and early postimplantation stages. https://doi.org/10.1289/EHP7349.
Sigma-Aldrich
Anti-PAX3 antibody produced in rabbit, affinity isolated antibody