MilliporeSigma
  • The mouse-equivalent of the human BDNF VAL66MET polymorphism increases dorsal hippocampal volume and does not interact with developmental ethanol exposure.
The mouse-equivalent of the human BDNF VAL66MET polymorphism increases dorsal hippocampal volume and does not interact with developmental ethanol exposure.
Alcohol (Fayetteville, N.Y.) (2020-04-01)
Clark W Bird, Megan J Barber, Jack Martin, Jacob J Mayfield, C Fernando Valenzuela
ABSTRACT
A relatively common polymorphism in the human brain-derived neurotrophic factor (BDNF) gene (Val66Met, which corresponds to Val68Met in mice) has been shown to modulate cognitive function and vulnerability to mental health disorders. This substitution impairs trafficking and activity-dependent release of BDNF. A number of studies with both humans and transgenic mice suggest that carriers of the Met allele have deficits in the structure and/or function of the hippocampal formation. Using a relatively new transgenic mouse model of this polymorphism, we recently demonstrated that it modulates the effects of developmental ethanol exposure in the hippocampus. Here, we further characterized the effect of this polymorphism on hippocampal morphology and its interaction with ethanol vapor exposure during the 2nd and 3rd trimester equivalents of human pregnancy. We found that BDNFmet/met mice have slightly larger hippocampal volumes than BDNFval/val mice. Ethanol vapor exposure during the 2nd and 3rd trimester equivalents of human pregnancy increased hippocampal volume in a single hippocampal subregion, the CA1 stratum radiatum. Ethanol exposure did not interact with BDNF genotype to affect volume in any hippocampal subregion. These results suggest that the Val66Met polymorphism does not reduce hippocampal size (i.e., it rather increases it slightly) or increase susceptibility to prenatal ethanol exposure-induced structural hippocampal damage during adulthood.
Sigma-Aldrich
Milli-Mark® FluoroPan Neuronal Marker - Alexa488 conjugated, Milli-Mark FluoroPan Neuronal Marker -Alexa488 conjugated is an antibody targeting the FluoroPan Neuronal Marker protein, validated for use in ICC & IF.