MilliporeSigma
  • Structure-Activity Relationship of Mono-Ion Complexes for Plasmid DNA Delivery by Muscular Injection.
Structure-Activity Relationship of Mono-Ion Complexes for Plasmid DNA Delivery by Muscular Injection.
Pharmaceutics (2021-01-13)
Amika Mori, Yuki Kobayashi, Kei Nirasawa, Yoichi Negishi, Shoichiro Asayama
ABSTRACT
The structure-activity relationship of mono-ion complexes (MICs) for plasmid DNA (pDNA) delivery by muscular injection is demonstrated. MICs were formed between pDNA and monocationic poly(ethylene glycol) (PEG) macromolecules. As monocationic PEGs, the ω-amide-pentylimidazolium (APe-Im) end-modified PEGs with a stable amide (Am) and hydrolytic ester (Es) bond, that is, APe-Im-Am-PEG and APe-Im-Es-PEG, respectively, are synthesized. The difference between the APe-Im-Am-PEG and APe-Im-Es-PEG was only a spacer structure between a terminal cation and a PEG chain. The resulting pDNA MICs with APe-Im-Am-PEG at a charge ratio (+/-) of 32 or 64 were more stable than those with APe-Im-Es-PEG in the presence of serum proteins. The highest gene expression by muscular injection was achieved using the APe-Im-Am-PEG/pDNA MIC at a charge ratio (+/-) of 32 with a smaller particle diameter of approximately 50 nm, as compared to that charge ratio of 64. Consequently, the pDNA MIC with the monocationic PEG with a stable amide spacer, as compared to a hydrolytic ester spacer, is considered to be suitable for the highest gene expression by muscular injection.
Sigma-Aldrich
1H-Imidazol-1-ylacetic acid, AldrichCPR