MilliporeSigma
  • Endothelial Mitochondrial Preprotein Translocase Tomm7-Rac1 Signaling Axis Dominates Cerebrovascular Network Homeostasis.
Endothelial Mitochondrial Preprotein Translocase Tomm7-Rac1 Signaling Axis Dominates Cerebrovascular Network Homeostasis.
Arteriosclerosis, thrombosis, and vascular biology (2018-10-26)
Dan Shi, Man Qi, Liping Zhou, Xiang Li, Le Ni, Changming Li, Tianyou Yuan, Yunqian Wang, Yongli Chen, Chaoyue Hu, Dandan Liang, Li Li, Yi Liu, Jun Li, Yi-Han Chen
ABSTRACT
Objective- Mitochondria are the important yet most underutilized target for cardio-cerebrovascular function integrity and disorders. The Tom (translocases of outer membrane) complex are the critical determinant of mitochondrial homeostasis for making organs acclimate physiological and pathological insults; however, their roles in the vascular system remain unknown. Approach and Results- A combination of studies in the vascular-specific transgenic zebrafish and genetically engineered mice was conducted. Vascular casting and imaging, endothelial angiogenesis, and mitochondrial protein import were performed to dissect potential mechanisms. A loss-of-function genetic screening in zebrafish identified that selective inactivation of the tomm7 (translocase of outer mitochondrial membrane 7) gene, which encodes a small subunit of the Tom complex, specially impaired cerebrovascular network formation. Ablation of the ortholog Tomm7 in mice recapitulated cerebrovascular abnormalities. Restoration of the cerebrovascular anomaly by an endothelial-specific transgenesis of tomm7 further indicated a defect in endothelial function. Mechanistically, Tomm7 deficit in endothelial cells induced an increased import of Rac1 (Ras-related C3 botulinum toxin substrate 1) protein into mitochondria and facilitated the mitochondrial Rac1-coupled redox signaling, which incurred angiogenic impairment that underlies cerebrovascular network malformation. Conclusions- Tomm7 drives brain angiogenesis and cerebrovascular network formation through modulating mitochondrial Rac1 signaling within the endothelium.
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.