MilliporeSigma
  • Synthesis and evaluation of curcumin analogues as potential thioredoxin reductase inhibitors.
Synthesis and evaluation of curcumin analogues as potential thioredoxin reductase inhibitors.
Bioorganic & medicinal chemistry (2008-08-06)
Xu Qiu, Zhong Liu, Wei-Yan Shao, Xing Liu, Da-Ping Jing, Yan-Jun Yu, Lin-Kun An, Shi-Liang Huang, Xian-Zhang Bu, Zhi-Shu Huang, Lian-Quan Gu
ABSTRACT
Series of curcumin derivatives were synthesized; the inhibitory activities on thioredoxin reductase (TrxR) of all analogues were evaluated by DTNB assay in vitro. It is found that most of the analogues can inhibit TrxR in the low micromolar range; Structure-activity relationship analysis reveals that analogues with furan moiety have excellent inhibitory effect on TrxR in an irreversible manner, indicating that the furan moiety may serve as a possible pharmacophore during the interaction of curcumin analogues with TrxR. The effect of selected curcuminoids on growth of different TrxR overexpressed cancer cell lines was also investigated and discussed.
Product Description
Supelco
Bisdemethoxycurcumin, analytical standard