MilliporeSigma
Application of carbamyl in structural optimization.
Bioorganic chemistry (2020-03-29)
Kuanglei Wang, Hongxi Zhu, Hongqian Zhao, Kun Zhang, Yongshou Tian
ABSTRACT
Carbamyl is considered a privileged structure in medicinal chemistry. It has a wide range of biological activities such as antimicrobial, anticancer, anti-epilepsy, for which the best evidence is a number of marketed carbamyl-containing drugs. Carbamyl is formed of primary amine and carbonyl moieties that act as hydrogen bond donors and hydrogen acceptors with residues of targets respectively, which are benefit for improving pharmacological activities. In other cases, the introduced carbamyl improves drug-like properties including oral bioavailability. In this review, we introduce the carbamyl-containing drugs and the application of carbamyl in structural optimization as a result of enhancing activities or/and drug-like properties.
Sigma-Aldrich
Imidafenacin hydrochloride, ≥95% (HPLC)