MilliporeSigma
  • The alpha-subunit of the colonic H+,K+-ATPase assembles with beta1-Na+,K+-ATPase in kidney and distal colon.
The alpha-subunit of the colonic H+,K+-ATPase assembles with beta1-Na+,K+-ATPase in kidney and distal colon.
The Journal of biological chemistry (1998-05-09)
J Codina, J T Delmas-Mata, T D DuBose
ABSTRACT
Previous experiments from our laboratory (Codina, J., Kone, B. C., Delmas-Mata, J. T., and DuBose, T. D., Jr. (1996) J. Biol. Chem. 271, 29759-29763) demonstrated that the alpha-subunit of the colonic H+, K+-ATPase (HKalpha2) requires coexpression with a beta-subunit to support H+/K+ transport in a heterologous expression system (Xenopus laevis oocytes). In these studies, HKalpha2 formed stable and functional alpha.beta complexes when coexpressed with either the rat beta1-subunit of the Na+,K+-ATPase or the beta-subunit of the gastric H+,K+-ATPase, suggesting that different beta-subunits may interact with HKalpha2. The present studies tested this hypothesis by development and application of a specific antibody against HKalpha2 peptide. Subsequently, immunoprecipitation experiments were performed to determine if HKalpha2 co-precipitates with the same beta-subunit in organs known to express HKalpha2 protein. The data demonstrate that HKalpha2 assembles with beta1-Na+,K+-ATPase in the renal medulla and in distal colon.
Sigma-Aldrich
Anti-Na+/K+ ATPase β-1 Antibody, clone C464.8, clone C464.8, Upstate®, from mouse