MilliporeSigma
  • Phase I Clinical Trial of Selinexor in Combination with Daunorubicin and Cytarabine in Previously Untreated Poor-Risk Acute Myeloid Leukemia.
Phase I Clinical Trial of Selinexor in Combination with Daunorubicin and Cytarabine in Previously Untreated Poor-Risk Acute Myeloid Leukemia.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-10-23)
Kendra Sweet, Rami Komrokji, Eric Padron, Christopher L Cubitt, Joel G Turner, Junmin Zhou, Alan F List, David A Sallman, Jana L Dawson, Daniel M Sullivan, Julio Chavez, Bijal D Shah, Jeffrey E Lancet
ABSTRACT
Induction chemotherapy results in complete remission (CR) rates of 20% to 50% among patients with poor-risk AML. Selinexor is an oral selective inhibitor of nuclear export with promising single-agent activity. By inhibiting the primary export protein, XPO1, selinexor localizes and activates tumor suppressor proteins in the nucleus and inhibits DNA damage repair, rationalizing combination with DNA-damaging agents. This was a single-arm phase I clinical trial of selinexor combined with cytarabine and daunorubicin (7+3). Dose escalation was selinexor alone (3+3) with an expansion at the MTD. Cohorts 1 and 2 received 60 and 80 mg orally, respectively, twice weekly during induction. Consolidation cycles (≤ 2) with selinexor at induction dose plus 5+2 were allowed for patients who achieved CR. MTD and recommended phase II dose of selinexor were the primary endpoints. Twenty-one patients with poor-risk AML were enrolled. All 21 patients were included in the safety evaluations and survival analyses (4 in each of 2 cohorts; 13 in the expansion); 8 (53%) of the 19 patients evaluable for response achieved CR/CRi. MTD was not reached. Selinexor 80 mg (orally, twice weekly) was used in the expansion phase. The most common grade 3/4 nonhematologic treatment-emergent adverse events were febrile neutropenia (67%), diarrhea (29%), hyponatremia (29%), and sepsis (14%). At median follow-up (28.9 months), 38% of patients were alive. Median overall survival was 10.3 months. Selinexor plus 7+3 is a safe regimen for patients with newly diagnosed poor-risk AML and warrants further investigation in a larger clinical trial.
Sigma-Aldrich
Anti-Topoisomerase II Antibody, clone KiS1, clone KiS1, Chemicon®, from mouse