MilliporeSigma
  • Metabolic functions of the human gut microbiota: the role of metalloenzymes.
Metabolic functions of the human gut microbiota: the role of metalloenzymes.
Natural product reports (2018-11-20)
Lauren J Rajakovich, Emily P Balskus
ABSTRACT
Covering: up to the end of 2017 The human body is composed of an equal number of human and microbial cells. While the microbial community inhabiting the human gastrointestinal tract plays an essential role in host health, these organisms have also been connected to various diseases. Yet, the gut microbial functions that modulate host biology are not well established. In this review, we describe metabolic functions of the human gut microbiota that involve metalloenzymes. These activities enable gut microbial colonization, mediate interactions with the host, and impact human health and disease. We highlight cases in which enzyme characterization has advanced our understanding of the gut microbiota and examples that illustrate the diverse ways in which metalloenzymes facilitate both essential and unique functions of this community. Finally, we analyze Human Microbiome Project sequencing datasets to assess the distribution of a prominent family of metalloenzymes in human-associated microbial communities, guiding future enzyme characterization efforts.
Sigma-Aldrich
4-Ethylphenol sodium sulfate