MilliporeSigma
  • Photoprotection by carotenoids during photosynthesis: motional dependence of intramolecular energy transfer.
Photoprotection by carotenoids during photosynthesis: motional dependence of intramolecular energy transfer.
Science (New York, N.Y.) (1982-05-28)
A L Moore, A Joy, R Tom, D Gust, T A Moore, R V Bensasson, E J Land
ABSTRACT
A new carotenoporphyrin has been prepared in which a synthetic carotenoid is joined to a tetraarylporphyrin through a flexible trimethylene linkage. This molecule exists primarily in an extended conformation with the carotenoid chromophore far from the porphyrin pi-electron system. In benzene solution, where large-amplitude molecular motions are rapid, the molecule can momentarily assume less stable conformations which favor triplet energy transfer, and quenching of the porphyrin triplet by the carotenoid is fast. In a polystyrene matrix or frozen glass such motions are slow, and energy transfer cannot compete with other pathways for depopulating the triplet state. These observations help establish the requirements for biological photoprotection.
Sigma-Aldrich
Pyropheophorbide a methyl ester, ≥93%