MilliporeSigma
  • Isolation and characterization of a marine bacterium Vibrio diabolicus strain L2-2 capable of biotransforming sulfonamides.
Isolation and characterization of a marine bacterium Vibrio diabolicus strain L2-2 capable of biotransforming sulfonamides.
Environmental research (2020-06-05)
Qiaoning Wang, Yaru Jiang, Hongdan Wang, Xianbo Chang, Min Lv, Lingxin Chen
ABSTRACT
Sulfonamides (SAs) have attracted much attention because of their high detection rates in natural water. In this study, a marine bacterium Vibrio diabolicus strain L2-2 was isolated which could metabolize 9 SAs to a different extent. Compared with SAs and their analogs, SAs with N-oxides of heterocyclic structure were easier to be transformed to their N4-acetylated metabolites or their isoxazole ring rearrangement isomers by strain L2-2. And, gene vdnatA and vdnatG were likely to be the key genes in SAs acetylation process, which might code Arylamine N-acetyltransferase. The biotransformation rates of sulfathiazole(STZ), sulfamonomethoxine(SMT), sulfadiazine(SDZ), sulfamethoxazole(SMX) and sulfisoxazole(SIX) could reach 29.39 ± 5.63, 24.97 ± 4.45, 79.41 ± 4.05, 64.64 ± 1.71, 32.82 ± 4.46% in 6 days, respectively. Besides, the overall optimal conditions for SAs biotransformation were less than 100 mg/L for total SAs in neutral or weakly alkaline medium with the salinity of 10-20‰ and additional nutrients like glucose, sucrose or glycerine. Furthermore, toxicity was demonstrated to be significantly reduced after biotransformation. Together, this study introduced a strategy to use V. diabolicus strain L2-2 to realize simultaneous removal and detoxification of multiple SAs in freshwater and seawater, and revealed SAs removal pathways and relevant molecular mechanism.
Sigma-Aldrich
4-aminophenyl methyl sulfone, AldrichCPR