MilliporeSigma
  • 4,5-di-O-caffeoylquinic acid methyl ester isolated from Lonicera japonica Thunb. targets the Keap1/Nrf2 pathway to attenuate H2O2-induced liver oxidative damage in HepG2 cells.
4,5-di-O-caffeoylquinic acid methyl ester isolated from Lonicera japonica Thunb. targets the Keap1/Nrf2 pathway to attenuate H2O2-induced liver oxidative damage in HepG2 cells.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2020-05-04)
Lingyun Xiao, Shu Liang, Lanlan Ge, Haoqiang Wan, Weigang Wu, Jia Fei, Shipin Wu, Boping Zhou, Xiaobin Zeng
ABSTRACT
4,5-di-O-caffeoylquinic acid methyl ester (4,5-CQME) is a caffeoylquinic acid (CQA) isolated from Lonicera japonica Thunb., a traditional Chinese medicine. To date, the biological activity of 4,5-CQME has not been fully investigated. The aim of the current study was to explore the anti-oxidative activity and the underlying mechanism of 4,5-CQME. MTT assay was used to evaluate the cytoprotective effect of 4,5-CQME. DCFH-DA was used as a fluorescence probe to detect intracellular ROS. The mitochondrial membrane potential was detected using the fluorescent probe JC-1. MDA and GSH levels were measured using MDA and GSH commercial kits, respectively. Apoptosis assay was performed using the Annexin V-FITC/PI method. The functional mechanism of 4,5-CQME was investigated by analyzing relative signaling pathways through immunofluorescent staining, quantitative PCR and western blot analysis. HepG2 cells were incubated with different concentrations of 4,5-CQME for 12 h before exposure to 500 μM H2O2 for 3 h. 4,5-CQME attenuated H2O2-induced oxidative damage and had a higher cytoprotective effect than 3-caffeoylquinic acid, 3-caffeoylquinic acid methyl ester, or 4,5-di-O-caffeoylquinic acid. 4,5-CQME also reduced ROS and MDA levels and rescued GSH depletion. Western blots demonstrated that 4,5-CQME decreased Bax/Bcl-2 and Bak levels. A mechanistic study confirmed that 4,5-CQME significantly suppressed H2O2-induced MAPKs phosphorylation but had little effect on MAPKs phosphorylation under normal conditions. By contrast, 4,5-CQME induced AKT phosphorylation in the presence or absence of H2O2. 4,5-CQME also regulated the Keap1/Nrf2 signaling pathway and enhanced both the mRNA and protein expressions of HO-1 and NQO1. The anti-oxidative effect of 4,5-CQME was greatly abolished by co-incubation with the Nrf2 inhibitor ML385 or PI3K inhibitor wortmannin. Taken together, these results showed that 4,5-CQME offered significant protection against H2O2-induced oxidative stress, and its effect was in part due to the modulation of the Keap1/Nrf2 pathway.
Sigma-Aldrich
4,5-Di-O-caffeoylquinic acid methyl ester, ≥90% (LC/MS-ELSD)