MilliporeSigma
  • THY1 is a candidate tumour suppressor gene with decreased expression in metastatic nasopharyngeal carcinoma.
THY1 is a candidate tumour suppressor gene with decreased expression in metastatic nasopharyngeal carcinoma.
Oncogene (2005-07-12)
Hong Lok Lung, Dhinoth Kumar Bangarusamy, Dan Xie, Arthur Kwok Leung Cheung, Yue Cheng, Mande Kuppusamy Kumaran, Lance Miller, Edison Tak-Bun Liu, Xin-Yuan Guan, Jonathan Shuntong Sham, Yan Fang, Liqiong Li, Nancy Wang, Alexey I Protopopov, Eugene R Zabarovsky, Sai Wah Tsao, Eric J Stanbridge, Maria Li Lung
ABSTRACT
Using oligonucleotide microarray analysis, THY1, mapping close to a previously defined 11q22-23 nasopharyngeal carcinoma (NPC) critical region was identified as showing consistent downregulated expression in the tumour segregants, as compared to their parental tumour-suppressing microcell hybrids (MCHs). Gene expression and protein analyses show that THY1 was not expressed in the NPC HONE1 recipient cells, tumour segregants, and other NPC cell lines; THY1 was exclusively expressed in the non-tumourigenic MCHs. The mechanism of THY1 gene inactivation in these cell lines was attributed to hypermethylation. Clinical study showed that in 65% of NPC specimens there was either downregulation or loss of THY1 gene expression. Using a tissue microarray and immunohistochemical staining, 44% of the NPC cases showed downregulated expression of THY1 and 9% lost THY1 expression. The frequency of THY1 downregulated expression in lymph node metastatic NPC was 63%, which was significantly higher than in the primary tumour (33%). After transfection of THY1 gene into HONE1 cells, a dramatic reduction of colony formation ability was observed. These findings suggest that THY1 is a good candidate tumour suppressor gene in NPC, which is significantly associated with lymph node metastases.