MilliporeSigma
  • Whole-cell bioconversion of naringenin to high added value hydroxylated compounds using Yarrowia lipolytica 2.2ab in surface and liquid cultures.
Whole-cell bioconversion of naringenin to high added value hydroxylated compounds using Yarrowia lipolytica 2.2ab in surface and liquid cultures.
Bioprocess and biosystems engineering (2020-03-08)
Christian Hernández-Guzmán, Arely Prado-Barragán, Miquel Gimeno, Angélica Román-Guerrero, Olga Miriam Rutiaga-Quiñones, Nuria Elizabeth Rocha Guzmán, Sergio Huerta-Ochoa
ABSTRACT
The bioconversion process of bioactive naringenin by whole-cells of Yarrowia lipolytica 2.2ab for the production of increased value-added compounds is successfully achieved in surface and liquid cultures. This approach is an alternative to the commercial production of these bioactive compounds from vegetable sources, which are limited due to their low concentrations and the complexity of the purification processes. The experimentation rendered seven value-added compounds in both surface and liquid bioconversion cultures. Some of the compounds produced have not been previously reported as products from the bioconversion processes, such as the case of ampelopsin. Biosynthetic pathways were suggested for the naringenin bioconversion using whole-cells of Y. lipolytica 2.2ab. Finally, the extracts obtained from the naringenin bioconversion in liquid cultures showed higher percentage of inhibition of DPPH· and ABTS· radicals up to 32.88 and 2.08 times, respectively, compared to commercial naringenin.
Product Description
Supelco
Aluminum crimp seal (without septum), silver aluminum,
pressure release
, diam. × opening 20 mm × 8 mm, pkg of × 100 ea