MilliporeSigma
  • Identification of Mycoplasmas using a fluorophore-free microarray and infrared chemical imaging (IRCI).
Identification of Mycoplasmas using a fluorophore-free microarray and infrared chemical imaging (IRCI).
Journal of microbiological methods (2011-06-23)
M M Mossoba, V Chizhikov, D V Volokhov, K Martinez-Diaz Milians, B Schoen, S F Al-Khaldi
ABSTRACT
A novel application of mid-infrared chemical imaging (IRCI) for the fluorophore-free detection and identification of mycoplasma species is reported for the first time. The PCR-amplified biotinylated targets hybridized to microarray probes were treated with streptavidin-gold nanoparticles followed by silver enhancement. This modification has the potential to expand the implementation of DNA microarray techniques in laboratories involved in the detection of cell substrates, other biological products, and clinical materials for the presence of mycoplasmas.
Sigma-Aldrich
Streptavidin−Gold from Streptomyces avidinii, 40 nm particle size