MilliporeSigma
  • Thiol-yne radical reaction mediated site-specific protein labeling via genetic incorporation of an alkynyl-L-lysine analogue.
Thiol-yne radical reaction mediated site-specific protein labeling via genetic incorporation of an alkynyl-L-lysine analogue.
Organic & biomolecular chemistry (2013-03-02)
Yiming Li, Man Pan, Yitong Li, Yichao Huang, Qingxiang Guo
ABSTRACT
Three alkyne-containing pyrrolysine derivatives were synthesized and genetically encoded into proteins by a mutant PylRS-tRNA pair with high efficiencies. With these alkyne handles, site-specific dual labeling of proteins can be achieved via a bioorthogonal thiol-yne ligation reaction.