MilliporeSigma
  • Palladium-catalyzed oxidative carbonylation of benzylic C-H bonds via nondirected C(sp3)-H activation.
Palladium-catalyzed oxidative carbonylation of benzylic C-H bonds via nondirected C(sp3)-H activation.
Journal of the American Chemical Society (2012-06-06)
Pan Xie, Yinjun Xie, Bo Qian, Han Zhou, Chungu Xia, Hanmin Huang
ABSTRACT
A new strategy for generating benzylpalladium reactive species from toluenes via nondirected C(sp(3))-H activation has been developed. This led to construction of an efficient Pd-catalyzed reaction protocol for the oxidative carboxylation of benzylic C-H bonds to form substituted 2-phenylacetic acid esters and derivatives from inexpensive, commercially available starting materials.
Sigma-Aldrich
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), 95%