MilliporeSigma
Interaction of JMJD6 with single-stranded RNA.
Proceedings of the National Academy of Sciences of the United States of America (2010-08-04)
Xia Hong, Jianye Zang, Janice White, Chao Wang, Cheol-Ho Pan, Rui Zhao, Robert C Murphy, Shaodong Dai, Peter Henson, John W Kappler, James Hagman, Gongyi Zhang
ABSTRACT
JMJD6 is a Jumonji C domain-containing hydroxylase. JMJD6 binds alpha-ketoglutarate and iron and has been characterized as either a histone arginine demethylase or U2AF65 lysyl hydroxylase. Here, we describe the structures of JMJD6 with and without alpha-ketoglutarate, which revealed a novel substrate binding groove and two positively charged surfaces. The structures also contain a stack of aromatic residues located near the active center. The side chain of one residue within this stack assumed different conformations in the two structures. Interestingly, JMJD6 bound efficiently to single-stranded RNA, but not to single-stranded DNA, double-stranded RNA, or double-stranded DNA. These structural features and truncation analysis of JMJD6 suggest that JMJD6 may bind and modify single-stand RNA rather than the previously reported peptide substrates.
Sigma-Aldrich
Papain Agarose from papaya latex, lyophilized powder, 90-150 units/mL packed gel