MilliporeSigma
  • Strong graphene oxide nanocomposites from aqueous hybrid liquid crystals.
Strong graphene oxide nanocomposites from aqueous hybrid liquid crystals.
Nature communications (2020-02-13)
Maruti Hegde, Lin Yang, Francesco Vita, Ryan J Fox, Renee van de Watering, Ben Norder, Ugo Lafont, Oriano Francescangeli, Louis A Madsen, Stephen J Picken, Edward T Samulski, Theo J Dingemans
ABSTRACT
Combining polymers with small amounts of stiff carbon-based nanofillers such as graphene or graphene oxide is expected to yield low-density nanocomposites with exceptional mechanical properties. However, such nanocomposites have remained elusive because of incompatibilities between fillers and polymers that are further compounded by processing difficulties. Here we report a water-based process to obtain highly reinforced nanocomposite films by simple mixing of two liquid crystalline solutions: a colloidal nematic phase comprised of graphene oxide platelets and a nematic phase formed by a rod-like high-performance aramid. Upon drying the resulting hybrid biaxial nematic phase, we obtain robust, structural nanocomposites reinforced with graphene oxide.
Sigma-Aldrich
Terephthaloyl chloride, ≥99%, flakes