MilliporeSigma
  • Long noncoding RNAs expressed in human hepatic stellate cells form networks with extracellular matrix proteins.
Long noncoding RNAs expressed in human hepatic stellate cells form networks with extracellular matrix proteins.
Genome medicine (2016-03-24)
Chan Zhou, Samuel R York, Jennifer Y Chen, Joshua V Pondick, Daniel L Motola, Raymond T Chung, Alan C Mullen
ABSTRACT
Hepatic fibrosis is the underlying cause of cirrhosis and liver failure in nearly every form of chronic liver disease, and hepatic stellate cells (HSCs) are the primary cell type responsible for fibrosis. Long noncoding RNAs (lncRNAs) are increasingly recognized as regulators of development and disease; however, little is known about their expression in human HSCs and their function in hepatic fibrosis. We performed RNA sequencing and ab initio assembly of RNA transcripts to define the lncRNAs expressed in human HSC myofibroblasts. We analyzed chromatin immunoprecipitation data and expression data to identify lncRNAs that were regulated by transforming growth factor beta (TGF-β) signaling, associated with super-enhancers and restricted in expression to HSCs compared with 43 human tissues and cell types. Co-expression network analyses were performed to discover functional modules of lncRNAs, and principle component analysis and K-mean clustering were used to compare lncRNA expression in HSCs with other myofibroblast cell types. We identified over 3600 lncRNAs that are expressed in human HSC myofibroblasts. Many are regulated by TGF-β, a major fibrotic signal, and form networks with genes encoding key components of the extracellular matrix (ECM), which is the substrate of the fibrotic scar. The lncRNAs directly regulated by TGF-β signaling are also enriched at super-enhancers. More than 400 of the lncRNAs identified in HSCs are uniquely expressed in HSCs compared with 43 other human tissues and cell types and HSC myofibroblasts demonstrate different patterns of lncRNA expression compared with myofibroblasts originating from other tissues. Co-expression analyses identified a subset of lncRNAs that are tightly linked to collagen genes and numerous proteins that regulate the ECM during formation of the fibrotic scar. Finally, we identified lncRNAs that are induced during progression of human liver disease. lncRNAs are likely key contributors to the formation and progression of fibrosis in human liver disease.
Sigma-Aldrich
Anti-PTPα Antibody, Upstate®, from rabbit