MilliporeSigma
  • ZC3H13 suppresses colorectal cancer proliferation and invasion via inactivating Ras-ERK signaling.
ZC3H13 suppresses colorectal cancer proliferation and invasion via inactivating Ras-ERK signaling.
Journal of cellular physiology (2018-10-13)
Dehua Zhu, Jianping Zhou, Jinbo Zhao, Guiyang Jiang, Xiupeng Zhang, Yong Zhang, Ming Dong
ABSTRACT
ZC3H13 is a canonical CCCH zinc finger protein, which harbors a somatic frame-shift mutation in colorectal cancer (CRC). However, its expression and biological function were still uncertain. In the current study, we found that ZC3H13 was served as a tumor suppressor in CRC cells, which decreased the expression of Snail, Cyclin D1, and Cyclin E1, and increased the expression of Occludin and Zo-1 through inactivating Ras-ERK signaling pathway. Furthermore, reduction of ZC3H13 associated with advanced TNM stage (p = 0.02), positive regional lymph node metastasis ( p = 0.01). Taken together, the current study indicated that ZC3H13 may be an upstream regulator of Ras-ERK signaling pathway and suppressed invasion and proliferation of CRC.
Sigma-Aldrich
Anti-ZC3H13 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution