MilliporeSigma
  • MKK7 transcription positively or negatively regulated by SP1 and KLF5 depends on HDAC4 activity in glioma.
MKK7 transcription positively or negatively regulated by SP1 and KLF5 depends on HDAC4 activity in glioma.
International journal of cancer (2019-04-10)
Yezhong Wang, Yong Xia, Kunhua Hu, Minling Zeng, Cheng Zhi, Miaoling Lai, Liqiang Wu, Sisi Liu, Shulian Zeng, Ziyan Huang, Shanshan Ma, Zhongmin Yuan
ABSTRACT
JNK activity has been implicated in the malignant proliferation, invasion and drug-resistance of glioma cells (GCs), but the molecular mechanisms underlying JNK activation are currently unknown. Here, we reported that MKK7, not MKK4, directly activates JNK in GCs and exerts oncogenic effects on tumor formation. Notably, MKK7 expression in glioma tissues was closely correlated with the grade of the glioma and JNK/c-Jun activation. Mechanistically, MKK7 transcription critically depends on the complexes formed by HDAC4 and the transcriptional factors SP1 and Krüppel-like factor-5 (KLF5), wherein HDAC4 directly deacetylates both SP1 and KLF5 and synergistically upregulates MKK7 transcription through two SP1 sites located on its promoter. In contrast, the increases in acetylated-SP1 and acetylated-KLF5 after HDAC4 inhibition switched to transcriptionally suppress MKK7. Selective inhibition of HDAC4 by LMK235, siRNAs or blockage of SP1 and KLF5 by the ectopic dominant-negative SP1 greatly reduced the malignant capacity of GCs. Furthermore, suppression of both MKK7 expression and JNK/c-Jun activities was involved in the tumor-growth inhibitory effects induced by LMK235 in U87-xenograft mice. Interestingly, HDAC4 is highly expressed in glioma tissues, and the rate of HDAC4 nuclear import is closely correlated with glioma grade, as well as with MKK7 expression. Collectively, these findings demonstrated that highly expressed MKK7 contributes to JNK/c-Jun signaling-mediated glioma formation. MKK7 transcription, regulated by SP1 and KLF5, critically depends on HDAC4 activity, and inhibition of HDAC4 presents a potential strategy for suppressing the oncogenic roles of MKK7/JNK/c-Jun signaling in GCs.
Sigma-Aldrich
M344, ≥98% (HPLC), powder
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
SAHA, ≥98% (HPLC)