MilliporeSigma
  • Determination of carbazole and halogenated carbazoles in human serum samples using GC-MS/MS.
Determination of carbazole and halogenated carbazoles in human serum samples using GC-MS/MS.
Ecotoxicology and environmental safety (2019-09-14)
Li Fang, Fengmei Qiu, Ying Li, Shizhong Wang, Josephine DeGuzman, Jun Wang, Jianwen She
ABSTRACT
Carbazole and halogenated carbazoles have been widely detected throughout the environment in soil, river deposits, and lake sediments. Human exposure to these compounds may occur through inhalation, drinking water, dietary intake and/or skin contact, and exposure levels in the body may be evaluated by measuring them in serum or blood. This paper reports the method development and validation for the analysis of carbazole and 11 halogenated carbazoles in human blood and/or serum samples. A small sample size of 100 μL of blood or serum was employed for the analysis. The samples were prepared through salting-out liquid-liquid extraction (LLE) by using hexane/ethyl acetate (4:1, v/v) as the extraction solvent and aqueous MgSO4 (37.5 wt%) as the salting-out regent, respectively. Sample analysis was performed using gas-chromatography (GC) coupled with a tandem mass spectrometer (MS/MS) in an electron impact (EI) mode. The developed method demonstrated low detection limits in the range of 0.02-0.27 ng/mL, intra-day accuracy ranging from 81.2% to 125%, and inter-day accuracy from 91.0% to 117%. The intra- and inter-day precisions, calculated by relative standard deviations (RSDs), were in the ranges of 1.0-16.0% and 1.8-16.4%, respectively. The developed method was applied to the analysis of 50 human serum samples collected from pregnant women in Southern California in 2012. Low concentrations of carbazole were measured in 18 samples, while halogenated carbazoles were not detected in any of the samples.
Sigma-Aldrich
3,6-Dibromocarbazole, 97%