MilliporeSigma
  • Resting-state and Vocabulary Tasks Distinctively Inform On Age-Related Differences in the Functional Brain Connectome.
Resting-state and Vocabulary Tasks Distinctively Inform On Age-Related Differences in the Functional Brain Connectome.
Language, cognition and neuroscience (2019-08-29)
Perrine Ferré, Yassine Benhajali, Jason Steffener, Yaakov Stern, Yves Joanette, Pierre Bellec
ABSTRACT
Most of the current knowledge about age-related differences in brain neurofunctional organization stems from neuroimaging studies using either a "resting state" paradigm, or cognitive tasks for which performance decreases with age. However, it remains to be known if comparable age-related differences are found when participants engage in cognitive activities for which performance is maintained with age, such as vocabulary knowledge tasks. A functional connectivity analysis was performed on 286 adults ranging from 18 to 80 years old, based either on a resting state paradigm or when engaged in vocabulary tasks. Notable increases in connectivity of regions of the language network were observed during task completion. Conversely, only age-related decreases were observed across the whole connectome during resting-state. While vocabulary accuracy increased with age, no interaction was found between functional connectivity, age and task accuracy or proxies of cognitive reserve, suggesting that older individuals typically benefits from semantic knowledge accumulated throughout one's life trajectory, without the need for compensatory mechanisms.