MilliporeSigma
  • LRRC8A potentiates temozolomide sensitivity in glioma cells via activating mitochondria-dependent apoptotic pathway.
LRRC8A potentiates temozolomide sensitivity in glioma cells via activating mitochondria-dependent apoptotic pathway.
Human cell (2018-11-15)
Chao Yang, Longshuang He, Gaofei Chen, Zeqian Ning, Zhibai Xia
ABSTRACT
Chloride (Cl-), a primary anion in the extracellular fluid, plays an important role in a variety of physiological and pathological processes, such as cell apoptosis and proliferation. However, the information about Cl- in cancer cell apoptosis and chemoresistance is poorly understood. In the present study, we found that temozolomide (TMZ) treatment led to a decrease in intracellular concentration of Cl- ([Cl-]i) in both U87 and TMZ-resistant U87/R glioma cells. The decrease in [Cl-]i was more noticeable in U87 cells than in U87/R cells. Moreover, the expression of LRRC8A was reduced in U87/R cells compared with U87 cells. LRRC8A downregulation inhibited TMZ, induced the decrease in [Cl-]i and abolished the difference of [Cl-]i between U87 cells and U87/R cells. Knockdown of LRRC8A using small interfering RNA attenuated TMZ-induced U87 cell growth inhibition and apoptosis, while overexpression of LRRC8A by adenoviral infection enhanced the effect of TMZ on U87 and U87/R cell viability and apoptosis. Furthermore, LRRC8A downregulation inhibited TMZ-induced mitochondria-dependent apoptosis, including elevated Bcl-2 expression, reduced Bax expression, cytochrome c release, and caspase nine and caspase three activation. On the contrary, upregulation of LRRC8A augmented the activation of mitochondria-dependent apoptotic pathway in U87 and U87/R cells. In conclusion, this study demonstrates that LRRC8A potentiates TMZ-induced glioma cell apoptosis via promoting mitochondria-dependent apoptosis, suggesting that LRRC8A can be represented as a novel target for drug resistance treatment in glioma cells.