MilliporeSigma
  • ZNF185 is a p63 target gene critical for epidermal differentiation and squamous cell carcinoma development.
ZNF185 is a p63 target gene critical for epidermal differentiation and squamous cell carcinoma development.
Oncogene (2018-10-20)
Artem Smirnov, Anna Maria Lena, Angela Cappello, Emanuele Panatta, Lucia Anemona, Simone Bischetti, Margherita Annicchiarico-Petruzzelli, Alessandro Mauriello, Gerry Melino, Eleonora Candi
ABSTRACT
Development and maintenance of healthy stratified epithelia require the coordination of complex transcriptional programmes. The transcription factor p63, a member of the p53 family, plays a crucial role in epithelial development and homeostasis. Analysis of the p63-dependent transcriptome indicated that one important aspect of p63 functions in epithelial development is the regulation of cell-cell and cell-matrix adhesion programmes. However, limited knowledge exists on the relevant cell-cell adhesion molecules involved in physiological epithelial formation. Similarly, limited data are available to understand if deregulation of the cell-cell adhesion programme is important in tumour formation. Here, using the epidermis as an experimental model with the RNA sequencing approach, we identify a novel p63-regulated gene induced during differentiation, ZNF185. ZNF185 is an actin-cytoskeleton-associated Lin-l 1, Isl-1 and Mec-3 (LIM) domain-containing protein, whose function is poorly known. We found that p63 binds to a specific enhancer region, promoting its expression to sustain epithelial differentiation. ZNF185 silencing strongly impaired keratinocyte differentiation according to gene array analysis. ZNF185 is detected at the cell-cell periphery where it physically interacts with E-cadherin, indicating that it is important to maintain epithelial integrity beyond its pro-differentiation role. Interestingly, poorly differentiated, including head and neck, cervical and oesophageal, squamous cell carcinomas display loss of ZNF185 expression. Together, these studies reinforce that p63 is a crucial gene for maintaining epithelial tissue integrity and support the deregulation of the cell-cell adhesion programme,which plays a critical role in carcinoma development.