MilliporeSigma
  • Physiological response of fish under variable acidic conditions: a molecular approach through the assessment of an eco-physiological marker in the brain.
Physiological response of fish under variable acidic conditions: a molecular approach through the assessment of an eco-physiological marker in the brain.
Environmental science and pollution research international (2019-06-15)
Amrita Mukherjee, Amiya Ranjan Bhowmick, Joyita Mukherjee, Mahammed Moniruzzaman
ABSTRACT
The current study demonstrates oxidative damage and associated neurotoxicity following pH stress in two freshwater carp Labeo rohita and Cirrhinus cirrhosus. Carp (n = 6, 3 replicates) were exposed to four different pH (5.5, 6, 7.5, and 8) against control (pH 6.8 ± 0.05) for 7 days. After completion of treatment, levels of enzymatic (superoxide dismutase [SOD], catalase [CAT], glutathione reductase [GRd]) and non-enzymatic antioxidants (malondialdehyde [MDA], glutathione [GSH]), brain neurological parameters (Na+-K+ATPase, acetylcholinesterase [AcHE], monoamine oxidase [MAO], and nitric oxide [NO]), xanthine oxidase (XO), heat shock proteins (HSP70 and HSP90), and transcription factor NFkB were measured in carp brain. Variation in the pH caused a significant alteration in the glutathione system (glutathione and glutathione reductase), SOD-CAT system, and stress marker malondialdehyde (MDA). Xanthine oxidase was also induced significantly after pH exposure. Brain neurological parameters (MAO, NO, AChE, and Na+-K+ATPase) were significantly reduced at each pH-treated carp group though inhibition was highest at lower acidic pH (5.5). Cirrhinus cirrhosus was more affected than that of Labeo rohita. Molecular chaperon HSP70 expression was induced in all pH-treated groups though such induction was more in acid-stressed fish. HSP90 was found to increase only in acid-stressed carp brain. Expression of NFkB was elevated significantly at each treatment group except for pH 7.5. Finally, both acidic and alkaline pH in the aquatic system was found to disturb oxidative balance in carp brain which ultimately affects the neurological activity in carp. However, acidic environment in the aquatic system was more detrimental than the alkaline system regarding oxidative damage and subsequent neurotoxicity in carp brain.