MilliporeSigma
  • Oral contraceptive use increases risk of inflammatory and coagulatory disorders in women with Polycystic Ovarian Syndrome: An observational study.
Oral contraceptive use increases risk of inflammatory and coagulatory disorders in women with Polycystic Ovarian Syndrome: An observational study.
Scientific reports (2019-07-17)
Saika Manzoor, Mohd Ashraf Ganie, Shajrul Amin, Zaffar A Shah, Imtiyaz A Bhat, S Douhath Yousuf, Humira Jeelani, Iram A Kawa, Qudsia Fatima, Fouzia Rashid
ABSTRACT
Polycystic ovarian syndrome (PCOS) is a multispectral disorder requiring lifelong management. Its pathophysiology is still being explored which makes its treatment options restrained. Present study explores impact of oral contraceptive mode of treatment on metabolic, hormonal, inflammation and coagulation profile of PCOS women. 50 subjects diagnosed with Rotterdam criteria receiving no drug treatment served as controls whereas 50 subjects receiving only OCPs (Ethinyl estradiol 0.03 mg, Levonorgestrel 0.15 mg) as a mode of treatment at least for six-months served as cases. Ferriman-Gallwey score and hormonal profile improved on OCP treatment. However, parameters like weight, Body mass index, waist-hip ratio, Oral glucose tolerance test, lipid profile, insulin, HOMA-IR, adiponectin, interleukin1β, visfatin, resistin, tissue factor, PT and APTT showed considerable derangements in OCP group. All above parameters are associated with the risk of diabetes mellitus, dyslipidemia, coronary vascular disease, cancers, hypercoagulable state, venous thromboembolism and thrombotic events. Long-term use of OCPs needs to be considered carefully for PCOS patients who are already burdened with associated risk factors. This study was conducted in a region where women do not have much access to high-end screening and diagnostic facilities that further exacerbates their clinical outcomes. Large scale, long-term studies need to be designed to further evaluate safety use of OCPs in PCOS women.