MilliporeSigma
  • Precise CCM1 gene correction and inactivation in patient-derived endothelial cells: Modeling Knudson's two-hit hypothesis in vitro.
Precise CCM1 gene correction and inactivation in patient-derived endothelial cells: Modeling Knudson's two-hit hypothesis in vitro.
Molecular genetics & genomic medicine (2019-05-28)
Stefanie Spiegler, Matthias Rath, Christiane D Much, Barbara S Sendtner, Ute Felbor
ABSTRACT
The CRISPR/Cas9 system has opened new perspectives to study the molecular basis of cerebral cavernous malformations (CCMs) in personalized disease models. However, precise genome editing in endothelial and other hard-to-transfect cells remains challenging. In a proof-of-principle study, we first isolated blood outgrowth endothelial cells (BOECs) from a CCM1 mutation carrier with multiple CCMs. In a CRISPR/Cas9 gene correction approach, a high-fidelity Cas9 variant was then transfected into patient-derived BOECs using a ribonucleoprotein complex and a single-strand DNA oligonucleotide. In addition, patient-specific CCM1 knockout clones were expanded after CRISPR/Cas9 gene inactivation. Deep sequencing demonstrated correction of the mutant allele in nearly 33% of all cells whereas no CRISPR/Cas9-induced mutations in predicted off-target loci were identified. Corrected BOECs could be cultured in cell mixtures but demonstrated impaired clonal survival. In contrast, CCM1-deficient BOECs displayed increased resistance to stress-induced apoptotic cell death and could be clonally expanded to high passages. When cultured together, CCM1-deficient BOECs largely replaced corrected as well as heterozygous BOECs. We here demonstrate that a non-viral CRISPR/Cas9 approach can not only be used for gene knockout but also for precise gene correction in hard-to-transfect endothelial cells (ECs). Comparing patient-derived isogenic CCM1+/+ , CCM1+/- , and CCM1-/- ECs, we show that the inactivation of the second allele results in clonal evolution of ECs lacking CCM1 which likely reflects the initiation phase of CCM genesis.