MilliporeSigma
  • Identification of Porphyromonas gingivalis lipopolysaccharide-binding proteins in human saliva.
Identification of Porphyromonas gingivalis lipopolysaccharide-binding proteins in human saliva.
Molecular immunology (2011-07-12)
Seulggie Choi, Jung Eun Baik, Jun Ho Jeon, Kun Cho, Deog-Gyu Seo, Kee-Yeon Kum, Cheol-Heui Yun, Seung Hyun Han
ABSTRACT
Porphyromonas gingivalis causes periodontal diseases and its lipopolysaccharide (LPS) is considered as a major virulence factor responsible for pathogenesis. Since initial recognition of P. gingivalis LPS (Pg.LPS) in the oral cavity might be crucial for the host response, we identified Pg.LPS-binding proteins (Pg.LPS-BPs) using Pg.LPS-immobilized beads and a high-resolution mass spectrometry. LPS purified from P. gingivalis was conjugated onto N-hydroxysuccinimidyl-Sepharose(®) 4 Fast Flow beads. Notably, Pg.LPS-conjugated beads could stimulate Toll-like receptor 2 (TLR2) as determined by a TLR2-depdendent reporter expression system using CHO/CD14/TLR2. In addition, the Pg.LPS-conjugated beads induced the production of inflammatory mediators such as nitric oxide and interferon-gamma-inducible protein-10 in the macrophage cell-line, RAW 264.7. These results imply that Pg.LPS retained its immunological properties during the conjugation process. Then, the Pg.LPS-conjugated beads were mixed with a pool of saliva obtained from nine human subjects to capture Pg.LPS-BPs and molecular identities were determined by LTQ-Orbitrap hybrid fourier transform mass spectrometry. Pg.LPS-BPs captured at high frequencies included alpha-amylase, cystatin, prolactin-inducible protein, lysozyme C, immunoglobulin components, serum albumin, lipocalin-1, and submaxillary gland androgen-regulated protein 3B. These proteins are known to be involved in bacterial adhesion and colonization, anti-microbial functions or modulation of immune responses.
Sigma-Aldrich
N-Hydroxysuccinimidyl-Sepharose 4 Fast Flow, matrix fast-flow highly cross-linked 4% beaded agarose