MilliporeSigma
  • SCA17, a novel autosomal dominant cerebellar ataxia caused by an expanded polyglutamine in TATA-binding protein.
SCA17, a novel autosomal dominant cerebellar ataxia caused by an expanded polyglutamine in TATA-binding protein.
Human molecular genetics (2001-07-13)
K Nakamura, S Y Jeong, T Uchihara, M Anno, K Nagashima, T Nagashima, S Ikeda, S Tsuji, I Kanazawa
ABSTRACT
Genetic etiologies of at least 20% of autosomal dominant cerebellar ataxias (ADCAs) have yet to be clarified. We identified a novel spinocerebellar ataxia (SCA) form in four Japanese pedigrees which is caused by an abnormal CAG expansion in the TATA-binding protein (TBP) gene, a general transcription initiation factor. Consequently, it has been added to the group of polyglutamine diseases. This abnormal expansion of glutamine tracts in TBP bears 47--55 repeats, whereas the normal repeat number ranges from 29 to 42. Immunocytochemical examination of a postmortem brain which carried 48 CAG repeats detected neuronal intranuclear inclusion bodies that stained with anti-ubiquitin antibody, anti-TBP antibody and with the 1C2 antibody that recognizes specifically expanded pathological polyglutamine tracts. We therefore propose that this new disease be called SCA17 (TBP disease).