MilliporeSigma
Characterization of adenosine receptors in bovine corneal endothelium.
Experimental eye research (2005-05-03)
Kah Y Tan-Allen, Xing Cai Sun, Joseph A Bonanno
ABSTRACT
Previous studies indicated that adenosine can increase [cAMP](i) and stimulate fluid transport by corneal endothelium. The purpose of this study was to determine which adenosine receptor subtype(s) are expressed and to examine their functional roles in modulating [cAMP](i), [Ca(2+)](i) and effects on Cl(-) permeability in corneal endothelium. We screened bovine corneal endothelium (BCE) for adenosine receptor subtypes by RT-PCR and immunoblotting, and examined the effects of pharmacological agents on adenosine stimulated Cl(-) transport, [cAMP](i) and [Ca(2+)](i). RT-PCR indicated the presence of A(1) and A(2b) adenosine receptors, while A(2a) and A(3) were negative. Western blot (WB) confirmed the presence of A(2b) ( approximately 50 kDa) and A(1) ( approximately 40 kDa) in fresh and cultured BCE. Ten micromolar adenosine increased [cAMP](i) by 2.7-fold over control and this was inhibited 66% by 10 microm alloxazine, a specific A(2b) blocker. A(1) activation with 1 micromN(6)-CPA (a specific A(1) agonist) or 100 nm adenosine decreased [cAMP](i) by 23 and 6%, respectively. Adenosine had no effect on [Ca(2+)](i) mobilization. Indirect immunofluorescence localized A(2b) receptors to the lateral membrane and A(1) to the apical surface in cultured BCE. Adenosine significantly increased apical Cl(-) permeability by 2.2 times and this effect was nearly abolished by DMPX (10 microm), a general A(2) blocker. Adenosine-induced membrane depolarization was also inhibited by 33% (n=6) in the presence of alloxazine. Bovine corneal endothelium expresses functional A(1) and A(2b) adenosine receptors. A(1), preferentially activated at <1 microm adenosine, acts to decrease [cAMP](i) and A(2b), activated at >1 microm adenosine, increase [cAMP](i).
Sigma-Aldrich
Anti-Adenosine A2a Receptor Antibody, serum, Chemicon®